55274-U
Supel™Genie C8 Online SPE Starter Kit
for use with SupelGenie Online SPE Cartridges, holder and 1 cartridge
About This Item
Recommended Products
material
stainless steel tube
product line
Supel™
composition
bed wt., 150 mg
packaging
pkg of 1 ea cartridge holder hardware and 1 cartridge
technique(s)
solid phase extraction (SPE): suitable
L × I.D.
20 mm × 4 mm
matrix active group
C8
application(s)
food and beverages
compatibility
for use with SupelGenie Online SPE Cartridges, holder and 1 cartridge
General description
Supel Genie Online SPE cartridges are designed for direct injection of untreated samples (e.g. water and urine) or processed samples (e.g. protein-precipitated plasma and serum), for analysis with liquid chromatography and mass spectrometry (LC/MS). The Supel Genie Online SPE cartridges enable online sample cleanup, concentration, and separation with excellent reproducibility and performance. Coupled with mass spectrometry for detection, the Supel Genie Online SPE cartridges provide a simple, automated method for sample analysis and are ideal for use in the LC/MS/MS analysis of biological fluids such as plasma and serum.
For first time use, a starter kit is required to obtain reusable universal hardware that will fit all Supel Genie Online SPE cartridges (P/N 55274-U).
LC instrumentation must be configured to accommodate Online SPE cartridges. Please see instrument setup diagram in datasheet. 55274-U
Please contact [email protected] for more information on setup and hardware.
Application
Features and Benefits
- Provides cleaner samples for longer column life, less instrument downtime, and more accurate and reproducible data
- Automation saves time, decreases costs, and eliminates human error
- Highly reproducible, consistent product performance
- C8 cartridges are ideal for reversed-phase extraction of hydrophobic or nonpolar to moderately polar compounds
- Starter kit comes with reusable universal hardware (fits any of the Supel Genie Online SPE cartridges) and 1 cartridge
- Cartridges also available in pack of 2 or 6
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service