Skip to Content
Merck
All Photos(1)

Key Documents

505773

Shutoff Valve, Low Pressure

blue Tefzel®, for use with 1/8 in. tubing

Synonym(s):

Tefzel® 1/8-in. 2-way shut-off valve

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 1,230.00

CZK 1,230.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CZK 1,230.00

About This Item

UNSPSC Code:
41115854

CZK 1,230.00


Please contact Customer Service for Availability

Request a Bulk Order

material

blue Tefzel®

description

manual shut-off valve

feature

positions 2

manufacturer/tradename

Upchurch Scientific, Inc P-783

parameter

600 psi max. pressure

fitting

1/4-28 female Upchurch in-line for 1/8 in. O.D. tubing

compatibility

for use with 1/8 in. tubing

Looking for similar products? Visit Product Comparison Guide

General description

  • Designed for lower pressure applications
  • Biocompatible, all polymer flow path

PEEK or Tefzel valve with 1/4-28 fittings, for pressures up to 1000 psi (70 kg/cm2). Flangeless nuts and ferrules included.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Tefzel is a registered trademark of E. I. du Pont de Nemours and Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service