Skip to Content
Merck
All Photos(1)

Key Documents

48608

Supelco

Dibromochloromethane solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 601

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/CHBr2Cl/c2-1(3)4/h1H

InChI key

GATVIKZLVQHOMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - Solvent

Flash Point(C)

9.7 °C - Solvent


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Zeng et al.
Environment international, 54, 134-140 (2013-03-05)
Toxicological studies showed that trihalomethanes (THMs), the most abundant classes of disinfection by-products (DBPs) in drinking water, impaired male reproductive health, but epidemiological evidence is limited and inconsistent. This study aimed to examine the associations of baseline blood THMs with
M S Christian et al.
International journal of toxicology, 20(4), 239-253 (2001-09-21)
Dibromoacetic acid (DBA) and bromodichloromethane (BDCM), by-products of chlorine disinfection of water, were provided in drinking water in range-finding reproductive/developmental toxicity studies (rats) and a developmental toxicity study (BDCM) in rabbits. Studies included absorption and biodisposition of DBA and BDCM
M N Kostopoulou et al.
Chemosphere, 40(5), 527-532 (2000-02-09)
An investigation into the occurrence of volatile organic compounds (VOCs) was conducted for a period of two years in the surface waters of Northern Greece. Samples from four rivers and five lakes were taken seasonally and analyzed for VOCs. The
M L Da Silva et al.
Journal of toxicology and environmental health. Part A, 60(5), 343-353 (2000-07-27)
The blood kinetics of trihalomethanes has recently been reported to differ between an oral administration of any single trihalomethane (0.25 mmol/kg) [THMs: chloroform, bromoform, bromodichloromethane (BDCM), dibromochloromethane (DBCM)] and a combined administration of 0.25 mmol/kg of each of the 4
J Calderón et al.
Gaceta sanitaria, 16(3), 241-243 (2002-06-12)
The presence of trihalomethanes in the water systems of several Spanish cities has been demonstrated. The aim of this study was to analyze trihalomethane concentrations in the water provided for human consumption in the city of Barcelona. We analyzed 151

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service