Skip to Content
Merck
All Photos(1)

Key Documents

48524

Supelco

1,4-Dichlorobenzene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein:
1680023
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

5.07 (vs air)

vapor pressure

1.03 mmHg ( 25 °C)

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

173 °C (lit.)

mp

52-54 °C (lit.)

density

1.241 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

SMILES string

Clc1ccc(Cl)cc1

InChI

1S/C6H4Cl2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

OCJBOOLMMGQPQU-UHFFFAOYSA-N

Gene Information

human ... CYP2A6(1548)
mouse ... Cyp2a5(13087)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael C McCarthy et al.
Environmental health perspectives, 117(5), 790-796 (2009-05-30)
Ambient measurements of hazardous air pollutants (air toxics) have been used to validate model-predicted concentrations of air toxics but have not been used to perform risk screening at the national level. We used ambient concentrations of routinely measured air toxics
John J Sillery et al.
Pediatric emergency care, 25(4), 252-254 (2009-04-17)
Hemolytic anemia and methemoglobinemia are well-known adverse effects that follow ingestion of naphthalene mothballs. They are only rarely reported in association with ingestion of paradichlorobenzene mothballs. An asymptomatic boy presented to our pediatric emergency department after ingesting paradichlorobenzene mothballs. Three
Kanako Watanabe et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 25(11), 1301-1305 (2009-11-13)
A very simple and sensitive method for the simultaneous analysis of naphthalene and p-dichlorobenzene in human whole blood and urine by headspace capillary gas chromatography-mass spectrometry (GC-MS) is presented. The advantages of the method were that as much as 1
Priscilla A Guerrero et al.
Journal of the Air & Waste Management Association (1995), 62(9), 1075-1084 (2012-10-02)
p-Dichlorobenzene (p-DCB) and naphthalene are classified as hazardous air pollutants and rank highly among chronic chemical hazards in US. residences. Sources of p-DCB and naphthalene include moth repellents and deodorizers typically used in closets, garment bags, and toilet bowls. Nearly
Wang Suhua et al.
Biological trace element research, 136(3), 320-336 (2009-11-06)
Paradichlorobenzene (pDCB) has been used as a space deodorant and moth repellant, as well as an intermediate in the chemical industry. Given its broad applications and high volatility, considerable concern exists regarding the adverse health effects of pDCB in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service