Skip to Content
Merck
All Photos(1)

Key Documents

47947

Supelco

Ethyl acetate solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

General description

Ethyl acetate can be obtained via esterification reaction between ethyl alcohol and acetic acid in the presence of sulfuric acid.[1][2] It can find applications as a solvent in the production of inks, varnishes, fat, nitrocellulose, etc. It can also be used as a cleaning agent in textile industry and as a flavoring agent for in food and pharmaceutical industries.[1] It can also serve as a substitute for methyl ethyl ketone in food industry.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aerobic oxidation of aqueous ethanol using heterogeneous gold catalysts: Efficient routes to acetic acid and ethyl acetate
J?rgensen B, et al.
J. Catal., 251, 332-337 (2007)
Stellman, MJ
Encyclopaedia of Occupational Health and Safety: Guides, indexes, directory (1998)
Yihua Liu et al.
Journal of food science, 78(2), T372-T376 (2013-01-16)
Due to the widespread use and potential toxicity of organophosphorous pesticides (OPs), multiresidue monitoring of OPs in camellia oil has become increasingly important. A simple, rapid, and effective matrix solid-phase dispersion extraction for the determination of 15 organophosphorous pesticides in
Kaushik Banerjee et al.
Journal of chromatography. A, 1270, 283-295 (2012-11-28)
A residue analysis method for the simultaneous estimation of 349 pesticides, 11 PCBs and 15 PAHs extracted from grape, pomegranate, okra, tomato and onion matrices, was established by using a gas chromatograph coupled to an electron impact ionization triple quadrupole
Zhi-Qi Yin et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 38(7), 1008-1013 (2013-07-16)
Fourteen compounds were isolated and purified from the ethyl acetate of the ethanol extract of Shiaria bambusicola by various chromatographic methods, and their structures were elucidated by spectral techniques and physicochemical properties as hypocrellin A (1), hypocrellin B (2), hypocrellin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service