Skip to Content
Merck
All Photos(1)

Key Documents

442799

Supelco

Tetrahydrothiophene

analytical standard

Synonym(s):

Tetramethylene sulfide, Thiolane, Thiophane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8S
CAS Number:
Molecular Weight:
88.17
Beilstein:
102392
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

18 mmHg ( 25 °C)

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.504 (lit.)

bp

119 °C (lit.)

mp

−96 °C (lit.)

density

1 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

C1CCSC1

InChI

1S/C4H8S/c1-2-4-5-3-1/h1-4H2

InChI key

RAOIDOHSFRTOEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O L Voronova et al.
Eksperimental'naia i klinicheskaia farmakologiia, 73(10), 37-39 (2011-01-25)
Beginning with the first hours of experiments, cisplatin evoked an increase of chromosomal aberrations in CBA/CaLac bone marrow cells. Significant increase of structural infringements of chromosomes due to chromatid breaks was revealed in metaphase plates of murine bone marrow preparations
The ganglionic blocking action of thiophanium derivatives.
L O RANDALL et al.
The Journal of pharmacology and experimental therapeutics, 97(1), 48-57 (1949-09-01)
V I Smol'iakova et al.
Eksperimental'naia i klinicheskaia farmakologiia, 73(8), 32-34 (2010-10-06)
Carbon tetrachloride-induced hepatitis in rats is accompanied by blood hyperviscosity syndrome development. A course intragastric administration of thiophane under these conditions prevents the increase in whole blood viscosity by normalizing the microrheological indices (deformability and aggregation of erythrocytes), which is
V I Smol'iakova et al.
Eksperimental'naia i klinicheskaia farmakologiia, 74(8), 37-40 (2012-01-12)
A hepatoprotective effect of thiophan was studied on the model of carbon tetrachloride-induced hepatitis in rats. Therapeutic administration of thiophan repairs the antitoxic function of liver, normalizes cytolysis marker activity, and improves the synthetic function of liver and the carbohydrate
James C Ritchie et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 28(1), 22-33 (2002-12-24)
Using purified enzyme preparations, we investigated the actions of angiotensin-converting enzyme, aminopeptidase N, and endopeptidase 24.11 on corticotropin-releasing factor (CRF). The effects of inhibition of these enzymes on CRF action in rat anterior pituitary cultures were also determined. Finally, specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service