Skip to Content
Merck
All Photos(1)

Key Documents

442268

Supelco

1-Hexadecene

analytical standard

Synonym(s):

Cetene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13CH=CH2
CAS Number:
Molecular Weight:
224.43
Beilstein:
1753401
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

>0.8 (vs air)

vapor pressure

<0.2 mmHg ( 20 °C)

description

This product has been discontinued. It has been replaced by product 52276.

autoignition temp.

464 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.441 (lit.)

bp

274 °C (lit.)

mp

3-5 °C (lit.)

density

0.783 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCC=C

InChI

1S/C16H32/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3H,1,4-16H2,2H3

InChI key

GQEZCXVZFLOKMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuyuki Kubo et al.
mBio, 6(5), e01305-e01315 (2015-09-17)
The cucumber anthracnose fungus Colletotrichum orbiculare forms specialized cells called appressoria for host penetration. We identified a gene, FAM1, encoding a novel peroxin protein that is essential for peroxisome biogenesis and that associates with Woronin bodies (WBs), dense-core vesicles found
Contact allergy to polyvinylpyrrolidone/hexadecene copolymer previously reported.
A C De Groot
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society, 7(4), 260-260 (1996-12-01)
M Tobler et al.
Contact dermatitis, 26(5), 299-303 (1992-05-01)
In continuation of our preceding studies, we disclose long-term experiments to test more extensively the protective power of the novel 4H Glove. 2 compounds encountered in modern electron microscopy technique, 1-hexadecene (1-HD) and 2-hydroxyethyl acrylate (2-HEA), which elicit allergic and/or
C Barba et al.
Meat science, 90(3), 697-700 (2011-11-22)
Solid phase microextraction (SPME) coupled with either gas chromatography-ionization flame detector (CG-FID) or multidimensional gas chromatography-mass spectrometry (MDGC-MS) was evaluated for its ability to detect volatile hydrocarbons produced during the irradiation of cooked ham. The chromatogram of an irradiated sample
Werner Gnatzy et al.
Arthropod structure & development, 44(5), 491-507 (2015-09-10)
The digger wasp species Ampulex compressa produces its venom in two branched gland tubules. They terminate in a short common duct, which is bifurcated at its proximal end. One leg is linked with the venom reservoir, the other one extends

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service