Skip to Content
Merck
All Photos(1)

Documents

41902

Supelco

EPA TO-14A Chlorinated Hydrocarbon Subset Mix

100 ppb in nitrogen, pkg of 110 L (pi-marked cylinder), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116105

grade

analytical standard

Agency

EPA TO-14

packaging

pkg of 110 L (pi-marked cylinder)

manufacturer/tradename

Scott Gas T1902-110

concentration

100 ppb in nitrogen

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

General description

EPA TO-14A Chlorinated Hydrocarbon Subset Mix [100 ppb in nitrogen, pkg of 110 L (pi-marked cylinder)] may be used as a chemical reference standard for the qualitative calibration of air monitoring and industrial hygiene equipment. These standards are available in both liquid solutions and gaseous blends, to accommodate calibration of today′s variety of air monitoring and analysis devices and applications. Each standard includes a Certificate of Analysis summarizing the testing data. Each standard solution is provided in a flame-sealed amber ampoule. Each gaseous standard is provided in a transportable cylinder for ease of use in either the laboratory or the field.
These high pressure, transportable gas calibration blends for US EPA Method TO-14 ensure that laboratories can perform Method TO-14 and Clean Air Act analyses at a fraction of their former costs. Scott TO-14 gas blends are packaged in SCOTTY HP high pressure, transportable, laboratory-size cylinders. Scott TO-14 gas calibration blends are traceable to NIST reference mixtures (NIST SRM 1804) and are certified by Scott Gases for superior stability and accuracy.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

19 analytes, 100 ppb each in nitrogen

Analyte

Description

    Carbon tetrachloride
    Chloroethane
    Chloroform
    Chloromethane
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    Hexachloro-1,3-butadiene
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    Vinyl chloride
    See All (19)

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service