Skip to Content
Merck
All Photos(1)

Key Documents

40152-U

Supelco

4-Chlorotoluene solution

5000 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C7H7Cl/c1-6-2-4-7(8)5-3-6/h2-5H,1H3

InChI key

NPDACUSDTOMAMK-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Nishio et al.
Applied microbiology and biotechnology, 55(3), 321-325 (2001-05-09)
p-Cymene monooxygenase (CMO) from Pseudomonas putida F1 consists of a hydroxylase (CymA1) and a reductase component (CymA2) which initiate pcymene (p-isopropyltoluene) catabolism by oxidation of the methyl group to p-isopropylbenzyl alcohol (p-cumic alcohol). To study the possible diverse range of
C Arunagiri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1747-1756 (2011-06-18)
The FT-IR and FT-Raman spectra of 2-bromo-4-chlorotoluene (2B4CT) molecule have been recorded in the region 4000-400 cm(-1) and 3500-50 cm(-1), respectively. Optimized geometrical structures, harmonic vibrational frequencies, intensities, reduced mass, force constants and depolarization ratio have been computed by the
Ya-Jun Liu et al.
Physical chemistry chemical physics : PCCP, 7(23), 3938-3942 (2005-12-07)
Equilibrium geometries and vibrational frequencies of the ground and some excited states of p-chlorotoluene were calculated by the complete active space self-consistent field (CASSCF) method. Multi-reference CASSCF second order perturbation theory (MSCASPT2) calculations were performed on the vertical excitation energies
Mannar R Maurya et al.
Dalton transactions (Cambridge, England : 2003), (32)(32), 4220-4232 (2008-08-07)
3-Formylsalicylic acid (Hfsal), covalently bound to chloromethylated polystyrene (PS) and cross-linked with 5% divinylbenzene reacts with d,l-alanine and l-isoleucine to give the Schiff-base tridentate ligands PS-H(2)fsal-d,l-Ala and PS-H(2)fsal-l-Ile, respectively. These anchored ligands upon reaction with VOSO(4) and Cu(CH(3)COO)(2).H(2)O form the
Eric R Strieter et al.
Journal of the American Chemical Society, 125(46), 13978-13980 (2003-11-13)
A comparative kinetic examination of catalyst systems based on several monophosphinobiaryl ligands is reported. The bulk of the phosphine ligand controls the catalytic activity and the rate of catalyst activation with the catalyst based on 2-dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl providing the greatest activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service