Skip to Content
Merck
All Photos(1)

Key Documents

40073

Supelco

Benzo[k]fluoranthene solution

certified reference material, 1000 μg/mL in acetone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
Beilstein:
1873745
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

packaging

ampule of 1 mL

concentration

1000 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

c1ccc2cc-3c(cc2c1)-c4cccc5cccc-3c45

InChI

1S/C20H12/c1-2-6-15-12-19-17-10-4-8-13-7-3-9-16(20(13)17)18(19)11-14(15)5-1/h1-12H

InChI key

HAXBIWFMXWRORI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erin E Bessette et al.
Drug metabolism and disposition: the biological fate of chemicals, 33(3), 312-320 (2004-12-04)
Polycyclic aromatic hydrocarbons (PAHs) and heavy metals are often environmental cocontaminants that could interact to alter PAH carcinogenicity. The heavy metal, arsenite, and the PAH, benzo[k]fluoranthene, were used as prototypes to investigate, in human HepG2 cells, mechanisms whereby the bioactivation
L Q Pan et al.
Environmental pollution (Barking, Essex : 1987), 141(3), 443-451 (2005-11-08)
The effects of benzo(a)pyrene (BaP), benzo(k)fluoranthene (BkF) and their mixture on antioxidant enzyme activities and lipid peroxidation (LPO) levels of haemolymph of scallop (Chlamys ferrari) were studied. The superoxide dismutase (SOD) activities of 0.5 microg/L and 1.0 microg/L were significantly
Tohru Saitoh et al.
Talanta, 79(2), 177-182 (2009-06-30)
A simple and rapid method for the highly sensitive determination of polycyclic aromatic hydrocarbons (PAHs) in water was developed. Benzo[a]pyrene, benzo[k]fluoranthene, perylene, and pyrene in water were concentrated into sodium dodecyl sulfate (SDS)-alumina admicelles. The collection was performed by adding
G Verrhiest et al.
Ecotoxicology (London, England), 10(6), 363-372 (2002-01-05)
Polycyclic Aromatic Hydrocarbons (PAHs) are ubiquitous pollutants of sediments. Sediment quality criteria often use toxicity data for individual PAHs. However, PAHs always occur in field sediments as a complex mixture of compounds. In this study, the toxicity of phenanthrene (P)
K L Willett et al.
Toxicology and applied pharmacology, 177(3), 264-271 (2001-12-26)
Certainpolynuclear aromatic hydrocarbons (PAHs) such as benzo[a]pyrene (BaP) induce CYP1A-dependent enzyme activities. Because PAHs are ubiquitous environmental contaminants, and some are aryl hydrocarbon agonists, CYP1A has been used as a biomarker for PAH exposure. However, PAHs exist in the environment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service