Skip to Content
Merck
All Photos(1)

Key Documents

38708

Supelco

Cereulide solution

20 μg/g in acetonitrile, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
CZK 7,030.00

CZK 7,030.00


Please contact Customer Service for Availability


Select a Size

Change View
0.5 ML
CZK 7,030.00

About This Item

Empirical Formula (Hill Notation):
C57H96N6O18
CAS Number:
Molecular Weight:
1153.40
MDL number:
NACRES:
NA.24

CZK 7,030.00


Please contact Customer Service for Availability

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g in acetonitrile

format

single component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.

The cyclic peptide Cereulide is a strong cytotoxin produced by some strains of Bacillus cereus and Bacillus megaterium. When ingested, it causes nausea and vomiting. It is both heat and acid stable and can be present mainly in stark-rich foods such as pasta or rice. This certified reference material together with its isotope labelled analogue 42354 is suitable to be used for ISO method 18465:2017 or similar LC-MS methods.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service