Skip to Content
Merck
All Photos(1)

Documents

33165-U

Supelco

Trifluoroacetic anhydride

Synonym(s):

Trifluoroacetic anhydride (TFAA), acetic anhydride, boron trifluoride, borontrifluoride, butanol, hexafluoroisopropanol, trimethylamine (TMA), TFAA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CO)2O
CAS Number:
Molecular Weight:
210.03
Beilstein:
746197
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for GC derivatization

vapor pressure

6.28 psi ( 20 °C)

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Esterifications

packaging

pkg of 10 × 1 mL (33165-U)

technique(s)

GC/MS: suitable (SIM)
gas chromatography (GC): suitable (ECD)

refractive index

n20/D 1.3 (lit.)

bp

39.5-40 °C (lit.)

mp

−65 °C (lit.)

density

1.511 g/mL at 20 °C (lit.)

SMILES string

FC(F)(F)C(=O)OC(=O)C(F)(F)F

InChI

1S/C4F6O3/c5-3(6,7)1(11)13-2(12)4(8,9)10

InChI key

QAEDZJGFFMLHHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for derivatization of alditols, aldoses, aryl amines, phenolic amines, primany amines, amines, tertiary aromatic amines, amino acids, amphetamine, beta blocker drugs, bile acids, ß-carboline, carbohydrates, desipramine, dopamine, glutathione, histidine, homovanillic, iso-homovanillic and dihydroxyphenyl acetic acids, hexosamines, n-hydroxylamines, hydroxy acids, p-hydroxyamphetamine, 4-hydroxycoumarins, n-hydroxy-2-fluorenylacetamide, p-hydroxynorephedrine, 5-hydroxytryptamine, indoles, Iodoamino acids, metanephrine, methylphenidate, monosaccharides, norepinephrine, normetanephrine, oligosaccharides, peptide amide, phenols, phentermine, phenylthiohydantoin amino acid, postaglandins F and F2α, propranolol, tetrahydropapaveroline, thiamine, N--trifluoroacetyl and vanillymandelic acid.

Other Notes

Reagent for butyl ester, cabethoxy, di(trifluoroacetyl), hexafluoroisopropyl ester, methyl ester, acetyl or trifluoroacetyl phenylthihydantoins, trifluoroacetate, trifluoroacetyl (TFA), trifluoroacetamide and trifluoroacetyl vanillin.

related product

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R G Smith et al.
Biomedical mass spectrometry, 4(3), 146-151 (1977-06-01)
Quantitative analyses of subnanomole quantities of the polyamines putrescine, cadaverine, spermidine and spermine by the gas chromatography mass spectrometry technique of selected ion monitoring are complicated by effects of the chromatographic column. These effects include sample retention by the column
G Lhoëst et al.
Biomedical mass spectrometry, 3(1), 21-27 (1976-02-01)
The biological implications, the reaction possibilities and the structure of different derivatives of N-hydroxy-2-fluorenylacetamide have been described.
GLC determination of propranolol, other beta-blocking drugs, and metabolites in biological fluids and tissues.
T Walle
Journal of pharmaceutical sciences, 63(12), 1885-1891 (1974-12-01)
J Gal et al.
Journal of pharmaceutical sciences, 66(6), 866-869 (1977-06-01)
A GLC-mass spectrometric assay for methylphenidate in biological fluids was developed using the ethyl ester homolog of the drug as the internal standard. The procedure has a lower level of sensitivity of 1.2 ng/ml and is based on GLC-mass spectrometic
A sensitive gas chromatographic procedure for the estimation of noradrenaline, dopamine and 5-hydroxytryptamine in rat brain.
I L Martin et al.
Biochemical pharmacology, 22(4), 521-533 (1973-02-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service