Skip to Content
Merck
All Photos(1)

Documents

33038

Supelco

HMDS+TMCS+Pyridine, 3:1:9 (Sylon HTP)

pkg of 20 × 1 mL

Synonym(s):

Chlorotrimethylsilane, Hexamethyldisilazane, Trimethylchlorosilane, Trimethylsilyl chloride, 1,1,1,3,3,3–Hexamethyldisilane+Τrimethylchlorosilane+Pyridine, Sylon HTP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

grade

for GC derivatization

description

3:1:9 (SYLON HTP)

composition

1,1,3,3,3-Hexamethyldisilazane, 22.9%
Pyridine, 69.4%
Trimethylchlorosilane, 7.7 % (w/v)

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 20 × 1 mL

Looking for similar products? Visit Product Comparison Guide

Application

HMDS+TMCS+Pyridine in the ratio of 3:1:9 may be used as the silylating agent, during multi-targeted profiling of the major phytohormones by gas chromatography-mass spectrometry (GC-MS).

Legal Information

Sylon is a trademark of Sigma-Aldrich Co. LLC
Sylon CT is a trademark of Sigma-Aldrich Co. LLC

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Claudia Birkemeyer et al.
Journal of chromatography. A, 993(1-2), 89-102 (2003-05-09)
In the present investigation we report selection of the N-methyl-N-(tert.-butydimethylsilyl)trifluoroacetamide (MTBSTFA) reagent as the most comprehensive derivatization protocol among 17 tested reactions covering trifluoroacetylation, pentafluorobenzylation, methylations, and trimethylsilylations. MTBSTFA allowed easy and robust tert.-butyldimethylsilyl derivatization of 1-aminocyclopropane-1-carboxylic acid, indole-3-acetic acid
T. Stoming et al.
Life Sciences, 12, 425-425 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service