Skip to Content
Merck
All Photos(1)

Key Documents

303102

Supelco

High Ethane Natural Gas Reference Standard

part volume 14 L, analytical standard

Synonym(s):

Natural Gas Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

description

high ethane

form

gas

manufacturer/tradename

Scott Gas TN900010-14

availability

not available in Canada, EU

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

  • High Ethane Natural Gas Reference Standard is a multi-component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors and in situ industrial stack analyzers.
  • A certificate of analysis is included.
  • Packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.

Analyte

Description

    Nitrogen 9 mol %
    Carbon dioxide .5 mol %
    Methane 64 mol %
    Ethane 12.5 mol %
    Propane 7 mol %
    2-Methylpropane 3 mol %
    Butane 3 mol %
    2-Methylbutane .5 mol %
    Pentane .5 mol %
    See All (9)

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1B - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service