Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
28886-U
SLB®-IL100 Capillary GC Column
L × I.D. 60 m × 0.25 mm, df 0.20 μm
Select a Size
CZK 20,650.00
List PriceCZK 41,300.00Save 50%Select a Size
About This Item
CZK 20,650.00
List PriceCZK 41,300.00Save 50%Recommended Products
material
fused silica
Agency
ASTM® D6526
parameter
≤25-230 °C temperature (isothermal or programmed)
Beta value
313
df
0.20 μm
technique(s)
gas chromatography (GC): suitable
L × I.D.
60 m × 0.25 mm
matrix active group
Non-bonded; 1,9-Di(3-vinylimidazolium)nonane bis(trifluoromethanesulfonyl)imide phase
application(s)
chemicals and industrial polymers
column type
capillary highly polar
Looking for similar products? Visit Product Comparison Guide
General description
USP Code: None
Phase:
- Non-bonded
- 1,9-Di(3-vinylimidazolium)nonane bis(trifluoromethylsulfonyl)imide
- Subambient to 230 °C (isothermal or programmed)
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Ionic liquid capillary GC columns enhance polar analyte inertness, addressing challenges in chromatography with improved stationary phases.
Chromatograms
suitable for GC-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
-
Are all the SLB Ionic Liquid Capillary Columns GC-MS compatible?
1 answer-
SLB-IL59, SLB-IL60, SLB-IL61, SLB-IL76, SLB-IL82, SLB-IL100 and SLB-IL111 Ionic Liquid Capillary Columns are all GC-MS compatible.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service