Skip to Content
Merck
All Photos(1)

Documents

27261-U

Supelco

Assembled screw cap with hole with PTFE/silicone septum

black polypropylene hole cap, thread for 8-425, PTFE/silicone, Top Hat (septa and closure designed to fit together tightly), for use with 2 mL vial, 4.6 mm opening, pkg of 100 ea

Synonym(s):

PTFE/Silicone Septum Screw Cap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

material

PTFE/silicone
black polypropylene hole cap

feature

closure type black polypropylene hole cap
pre-assembled
Top Hat (septa and closure designed to fit together tightly)

packaging

pkg of 100 ea

fitting

thread for 8-425

compatibility

for use with 2 mL vial, 4.6 mm opening

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

The septa and closure was designed to provide a posiive physical lock to assure that preassembled components arrive intact.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service