Skip to Content
Merck
All Photos(1)

Documents

27020

Supelco

Assembled screw cap with hole with PTFE/silicone septum

black polypropylene hole cap, thread for 18-400, PTFE/silicone, septum thickness 1.5 mm, for use with 15 mL vial, pkg of 100 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

PTFE/silicone
black polypropylene hole cap

feature

closure type black polypropylene hole cap

packaging

pkg of 100 ea

septum thickness

1.5 mm

fitting

thread for 18-400

compatibility

for use with 15 mL vial

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

It may be used to cut and store fresh plant material for collecting volatiles in solid phase microextraction (SPME) technique. It may also be used in liquid chromatography–tandem mass spectrometry (LC–MS/MS) method for determining bisphenol A, 4-t-octylphenol and 4-nonylphenol in human blood serum.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bisphenol A, 4-t-octylphenol, and 4-nonylphenol determination in serum by Hybrid Solid Phase Extraction?Precipitation Technology technique tailored to liquid chromatography?tandem mass spectrometry.
Asimakopoulos, Alexandros G., and Nikolaos S. Thomaidis.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 986, 85-93 (2015)
Electroantennographic responses of the small chestnut weevil Curculio sayi (Coleoptera: Curculionidae) to volatile organic compounds identified from chestnut reproductive plant tissue.
Keesey, Ian W., et al.
Environmental Entomology, 41.4, 933-940 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service