Key Documents
25806
Supelco
"T"-Type Gas Line Filter
brass, 1/4 in. male
Select a Size
1 EA
CZK 4,660.00
CZK 4,660.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
CZK 4,660.00
About This Item
UNSPSC Code:
41112500
CZK 4,660.00
Please contact Customer Service for Availability
Request a Bulk Order
material
brass
fitting
1/4 in. male
General description
This gas line filter contains a replaceable 7 μm sintered stainless steel frit, and will remove dust and small particles that could otherwise foul downstream needle valves. Nuts and ferrules included.
Choose from one of the most recent versions:
Certificates of Analysis (COA)