Skip to Content
Merck

Skip To

22706

SWAGELOK® Connector to Female NPT

SWAGELOK® 400-7-2, stainless steel, 1/4 in. SWAGELOK, 1/8 in. female NPT

Synonym(s):

Swagelok® 400-7-2,SS CONNECTOR 1/4TX1/8P EA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
872,00 CZK

About This Item

eCl@ss:
32035302
UNSPSC Code:
41115717
NACRES:
SB.54

872,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

SWAGELOK® 400-7-2

technique(s)

gas chromatography (GC): suitable using :Suitable

fitting

1/4 in. SWAGELOK, 1/8 in. female NPT

General description

Nuts and ferrules included.

Legal Information

SWAGELOK is a registered trademark of Swagelok Company

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2270821978-U21519
description

SWAGELOK® 400-7-2, stainless steel, 1/4 in. SWAGELOK, 1/8 in. female NPT

description

SWAGELOK® 400-7-4, stainless steel, 1/4 in. SWAGELOK, 1/4 in. female NPT

description

SWAGELOK® 200-7-4, brass, 1/8 in. SWAGELOK, 1/4 in. female NPT

description

brass, 1/4 in. SWAGELOK, 1/4 in. male NPT, 400-1-4

fitting

1/4 in. SWAGELOK, 1/8 in. female NPT

fitting

1/4 in. SWAGELOK, 1/4 in. female NPT

fitting

1/4 in. female NPT, 1/8 in. SWAGELOK

fitting

1/4 in. SWAGELOK, 1/4 in. male NPT

packaging

pkg of 1 ea

packaging

pkg of 1 ea

packaging

pkg of 2 ea

packaging

pkg of 2 ea

manufacturer/tradename

SWAGELOK® 400-7-2

manufacturer/tradename

SWAGELOK® 400-7-4

manufacturer/tradename

SWAGELOK® 200-7-4

manufacturer/tradename

400-1-4

material

stainless steel

material

stainless steel

material

brass

material

brass


Still not finding the right product?

Explore all of our products under Swagelok® Connector to Female NPT




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters