Skip to Content
Merck
All Photos(1)

Documents

22137-U

Swagelok® Plug

Swagelok® 100-P, stainless steel, 1/16 in. Swagelok

Synonym(s):

Swagelok® 100P,SS PLUGS 1/16"EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 100-P

fitting

1/16 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

Application

Swagelok® plugs maybe used for closing the collection tubes, in combined capillary gas chromatography and mass spectrometry used for the identification of ocimene in alfalfa flower aroma.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of ocimene in alfalfa flower aroma by combined GC-mass spectrometry.
Loper GM
Crop Science, 11(1), 61-63 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service