Skip to Content
Merck

Skip To

22015

SWAGELOK® Union

SWAGELOK® 200-6, brass, 1/8 in.

Synonym(s):

Swagelok® 200-6,BRASS UNION 1/8" PK2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2 ea
Please contact Customer Service for Availability
636,00 CZK

About This Item

UNSPSC Code:
41115717
NACRES:
SB.54

636,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

brass

packaging

pkg of 2 ea

manufacturer/tradename

SWAGELOK® 200-6

technique(s)

gas chromatography (GC): suitable using :Suitable

fitting

1/8 in.

General description

Nut and ferrules included.
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.

Application

Brass Swagelok® union maybe used in determination of n-butylated trialkyllead compounds by gas chromatography with microwave plasma emission detection.

Legal Information

SWAGELOK is a registered trademark of Swagelok Company

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
22030-U2200425828
description

SWAGELOK® 200-6, brass, 1/8 in.

description

SWAGELOK® 400-6, stainless steel, 1/4 in.

description

SWAGELOK® 400-6, brass, 1/4 in.

description

SWAGELOK® 810-6, stainless steel, 1/2 in.

manufacturer/tradename

SWAGELOK® 200-6

manufacturer/tradename

SWAGELOK® 400-6

manufacturer/tradename

SWAGELOK® 400-6

manufacturer/tradename

SWAGELOK® 810-6

fitting

1/8 in.

fitting

1/4 in.

fitting

1/4 in.

fitting

1/2 in.

packaging

pkg of 2 ea

packaging

pkg of 1 ea

packaging

pkg of 2 ea

packaging

pkg of 1 ea

material

brass

material

stainless steel

material

brass

material

stainless steel


Still not finding the right product?

Explore all of our products under Swagelok® Union




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters