Skip to Content
Merck
All Photos(1)

Key Documents

21981-U

Swagelok® Bulkhead Union

Swagelok® 200-61, brass, 1/8 in.

Synonym(s):

Swagelok® 200-61, BRASS BULKHEAD UNION 1/8", EA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 780.00

CZK 780.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CZK 780.00

About This Item

UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54

CZK 780.00


Please contact Customer Service for Availability

Request a Bulk Order

material

brass

description

bulkhead design yes

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 200-61

fitting

1/8 in.

compatibility

for use with 1/8 in. tubing

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Swagelok® Bulkhead union may be used in constructing a solid-phase microextraction (SPME) fibre sampling port by placing the septum on the outer nut.[1]

Legal Information

Swagelok is a registered trademark of Swagelok Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole C Bouvier-Brown et al.
Journal of chromatography. A, 1161(1-2), 113-120 (2007-07-06)
Biogenic terpenes play important roles in ecosystem functioning and atmospheric chemistry. Some of these compounds are semi-volatile and highly reactive, such as sesquiterpenes and oxygenated terpenes, and are thus difficult to quantify using traditional air sampling and analysis methods. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service