Skip to Content
Merck
All Photos(1)

Key Documents

21965-U

SGE Gas Tight Syringes, Luer Lock

5MDR-LL-GT, volume 5 mL, needle size (not included)

Synonym(s):

SYRINGE SGE 5ML GAS TIGHT LUER LOCK, EA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 3,530.00

CZK 3,530.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CZK 3,530.00

About This Item

UNSPSC Code:
41122001
eCl@ss:
32070500
NACRES:
NB.03

CZK 3,530.00


Please contact Customer Service for Availability

Request a Bulk Order

material

glass syringe

needle size

(not included)

feature

needle type Luer lock

packaging

pkg of 1 ea

manufacturer/tradename

SGE 008760

volume

5 mL

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. what pressure range can this syringe handle? and what happends when this is exceeded?

    1 answer
    1. The pressure ratings are not determined for this product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service