Skip to Content
Merck
All Photos(1)

Key Documents

21006

Supelco

GC Stationary Phase

phase Apiezon L, bottle of 25 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717

form

solid

packaging

bottle of 25 g

parameter

50-300 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix active group

Apiezon L phase

solubility

chloroform, toluene: soluble

column type

packed GC

General description

GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients.[1] The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation and peak tailing are a few parameters which are considered while choosing a stationary phase.[2] Apiezon L is basically purified hydrocarbon grease which is suitable as a liquid phase for GC analysis. It is considered as one of the most selective phase for Polychlorinated Biphenyls (PCB) congeners.[3]
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Application

Apiezon L may be used as stationary phase to determine the retention indices of benzene and benzene derivatives on the basis of their molecular structure using GC equipped with a thermal conductivity detector.[4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitchell D. Erickson
Analytical Chemistry of PCBs, 250-250 (1997)
Calculation of retention indices for benzene and benzene derivatives on the basis of molecular structure.
Cook, Lawrence E., and Frank M. Raushel.
Journal of Chromatography A, 65, 556-559 (1972)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service