Skip to Content
Merck
All Photos(3)

Key Documents

20011-U

Supelco

Tenax® GR Thermal Desorption Tube

greener alternative

stainless steel, O.D. x I.D. x L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
CZK 30,100.00

CZK 30,100.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 EA
CZK 30,100.00

About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

CZK 30,100.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Tenax® GR, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea

material

stainless steel TD tube

description

Sealed with TDS³ Storage Container

feature

preconditioned

packaging

pkg of 10 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Tenax GR graphitized porous polymer

suitability

suitable for (extends range of Tenax GR)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Legal Information

Tenax is a registered trademark of Buchem B.V.

comparable product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service