12495-U
Metal Packed GC Column (General Configuration)
phase SP®-1000, 0.1%, matrix 80/100 Carbopack C support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
About This Item
Recommended Products
material
stainless steel column
parameter
0-225 °C temperature (isothermal or programmed)
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
matrix
80/100 Carbopack C support
matrix active group
SP®-1000 phase, 0.1%
column type
packed GC
compatibility
for use with General Configuration
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Kit Components Also Available Separately
- 11820Coated GC Packing, phase SP®-1000, 0.1%, matrix 80/100 Carbopack C support, bottle of 15 gSDS
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service