Skip to Content
Merck
All Photos(4)

Key Documents

ZRB2837

Sigma-Aldrich

Anti-LGR5/GPR49 Antibody, clone 1I17 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

G-protein coupled receptor 67, G-protein coupled receptor HG38, GPR49, GPR67, Leucine-rich repeat-containing G-protein coupled receptor 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I17, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 100 kDa
observed mol wt ~120 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... LGR5(8549)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1I17 is a ZooMAb® recombinant rabbit monoclonal antibody that detects LGR5/GPR49. It targets an epitope within 25 amino acids from the extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the extracellular domain of human LGR5/GPR49.

Application

Quality Control Testing

Evaluated by Western Blotting in HL-60 cell lysate.

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected LGR5/GPR49 in HL-60 cell lysate.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound LGR5/GPR49 peptide with at least ten thousand-fold (10,000X) higher than with non-specific control peptide.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected LGR5/GPR49 in human colon tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected LGR5/GPR4 in SH-SY5Y cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Leucine-rich repeat-containing G-protein coupled receptor 5 (UniProt: O75473; also known as G-protein coupled receptor 49, G-protein coupled receptor 67, G-protein coupled receptor HG38) is encoded by the LGR5 (also known as GPR49, GPR67) gene (Gene ID: 8549) in human. It is a G-protein coupled receptor (GPCR) characterized by an extracellular domain containing multiple leucine-rich repeat (LRR) sequences, critical for protein-protein interactions. The structure also includes a seven-transmembrane domain typical of GPCRs. LGR5/GPR49 is a receptor for R-spondins that potentiates the Wnt signaling pathway, a key regulator of cell proliferation, differentiation, and migration, stem cell renewal and growth. This receptor is crucial in maintaining the stem cell population in various tissues, particularly in the intestinal crypts where it supports rapid turnover and regeneration. Overexpression of LGR5/GPR49 has been observed in various cancer types, where it is associated with tumor growth, metastasis, and poor prognosis. It is a potential marker for identifying cancer stem cells within tumors, which are implicated in resistance to conventional therapies and in the recurrence of cancer.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service