Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2163

Sigma-Aldrich

Anti-PDH kinase 1 Antibody, clone 1C7-H1 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:2.7.11.2, Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 1, mitochondrial, Pyruvate dehydrogenase kinase isoform 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1C7-H1
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 49.24 kDa
observed mol wt ~44 kDa

purified by

using Protein A

species reactivity

mouse, human, rat

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PDK1(5163)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C7-H1 ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Pyruvate dehydrogenase kinase 1 (PDH kinase 1). It targets an epitope within 15 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Pyruvate dehydrogenase kinase 1 (PDH kinase 1).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected PDH kinase 1 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PDH kinase 1 in lysates from RAW264.7 and C6 cells.

Affinity Binding Assay: A representative lot of this antibody bound PDH kinase 1 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected PDH kinase 1 in HeLa cells.

Flow Cytometry Analysis: 1 µg from a representative lot detected PDH kinase 1 in one million Hela cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PDH kinase 1 in Human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 1, mitochondrial (UniProt: Q15118; also known as EC:2.7.11.2, Pyruvate dehydrogenase kinase isoform 1, PDH kinase 1) is encoded by the PDK1 (also known as PDHK1) gene (Gene ID: 5163) in human. PDH kinase 1 is a mitochondrial matrix enzyme that is synthesized with a transit peptide (aa 1-28), which is subsequently cleaved off. It is expressed predominantly in the heart and is detected at lower levels in liver, skeletal muscle, and pancreas. It plays a key role in regulation of glucose and fatty acid metabolism and homeostasis via phosphorylation of the pyruvate dehydrogenase subunits PDHA1 and PDHA2. This inhibits pyruvate dehydrogenase activity, and thereby regulates metabolite flux through the TCA cycle, down-regulates aerobic respiration and inhibits the formation of acetyl-CoA from pyruvate. It is also reported to play an important role in cellular responses to hypoxia and is shown to be important for cell proliferation under hypoxic conditions. It also protects cells against apoptosis in response to hypoxia and oxidative stress. Its levels are up-regulated via the HIF1 signaling in response to hypoxia. Exposure of cancer cells to severe hypoxia is reported to induce translocation of AKT to the mitochondrion, which leads to AKT-mediated phosphorylation of PDH kinase 1 at threonine 346 that supports tumor cell survival and proliferation during hypoxia. PDH kinase 1 is phosphorylated by constitutively activated Abl1, FGFR1, FLT3 and JAK2. Its phosphorylation at tyrosine 243 and 244 strongly increases its kinase activity, while phosphorylation at tyrosine 136 has reduced effect. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonal. (Ref.: Chae YC., et al. (2016). Cancer Cell. 30(2); 257-272; McFate, T., et al. (2008). J. Biol. Chem. 283(33); 22700-22708; Gudi, R., et al. (1995). J. Biol. Chem. 270(48); 28989-28994).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service