Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1786

Sigma-Aldrich

Anti-NDRG1 Antibody, clone 1C11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CZK 5,200.00
4 X 25 μL
CZK 10,600.00

CZK 5,200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
CZK 5,200.00
4 X 25 μL
CZK 10,600.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

CZK 5,200.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C11, recombinant monoclonal

description

1C11 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 42.84 kDa
observed mol wt ~48 kDa

purified by

using Protein A

species reactivity

human, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NDRG1(10397)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C11 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects NDRG1. It targets an epitope within 15 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of Human NDRG1.

Application

Quality Control Testing

Evaluated by Western Blotting in COS-7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected NDRG1 in COS-7 cell lysate.

Tested applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected NDRG1 in human kidney and human prostate tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound NDRG1 peptide with a KD of 3.9 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Protein NDRG1 (UniProt: Q92597; also known as Differentiation-related gene 1 protein, DRG-1, N-myc downstream-regulated gene 1 protein, Nickel-specific induction protein Cap43, Reducing agents and tunicamycin-responsive protein, RTP) is encoded by the NDRG1 (also known as CAP43, DRG1, RTP) gene (Gene ID: 10397) in human. NDRG1 is a stress-responsive protein that is induced under a wide variety of stress and cell growth-regulatory conditions. It is ubiquitously expressed but the most prominent expression is observed in placental membranes, prostate, kidney, small intestine, and ovary tissues. It is mainly a cytoplasmic protein, but is differentially localized to other regions. NDRG1 is reported to be up-regulated by cell differentiation signals in various cancer cell lines and also suppresses tumor metastasis. It is reported to play different roles in metastatic versus non-metastatic cells. It suppresses cell growth and proliferation in the metastatic H1299-NDRG1 cells, but does not have any effect on the non-metastatic DLD-1-NDRG1 cells. It is induced about 20-fold during in vitro differentiation of the colon carcinoma cell lines HT-29-D4 and Caco-2 cells. NDRG1 is shown to be essential, but not sufficient for p53/TP53-mediated caspase activation and apoptosis. It is also shown to have a role in cell trafficking of Schwann cells and is necessary for the maintenance and development of the peripheral nerve myelin sheath. NDRG1 is phosphorylated by several protein kinases, including PKA, PKC, and CaMK II. It is also phosphorylated by SGK1 on many serine and threonine residues in the C-terminal region, which is temporally and spatially controlled during the cell cycle. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.(Ref.: (Ref.: McCaig, C., et al. (2011). Biochem. Biophys. Res. Commun. 411(2); 227-234; Ellen, TP., et al. (2008). Carcinogenesis. 29(1); 2-8).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service