Skip to Content
Merck
All Photos(18)

Documents

ZRB1581

Sigma-Aldrich

Anti-p14ARF Antibody, clone 4F19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ARF, Alternative reading frame, Cyclin-dependent kinase inhibitor 2A, Tumor suppressor ARF, p14ARF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F19, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 13.9 kDa
observed mol wt ~38 kDa (GST-tagged recombinant protein)

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CDKN2A(1029)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 4F19 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects p14ARF.

Immunogen

GST/His-tagged full-length human recombinant Tumor suppressor ARF

Application

Anti-p14ARF, clone 4F19 ZooMAb, Cat. No. ZRB1581, is a recombinant Rabbit monoclonal antibody that specifically targets p14ARF and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected p14ARF in HeLa, A431, and NIH 3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected p14ARF in human testis tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound human recombinant p14ARF with a KD of 2.0 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Tumor suppressor ARF (UniProt: Q8N726; also known as Alternative reading frame, ARF, Cyclin-dependent kinase inhibitor 2A, p14ARF) is encoded by the CDKN2A (also known as CDKN2, MLM) gene (Gene ID: 1029) in human. p14ARF serves as a tumor suppressor and is a key regulator of cellular proliferation. It is shown to be frequently inactivated in human cancers. It binds to MDM2 and blocks its nucleocytoplasmic shuttling by sequestering it in the nucleolus. This inhibits the oncogenic action of MDM2 by blocking MDM2-induced degradation of p53 and enhances p53-dependent transactivation and apoptosis. Also induces G2 arrest and apoptosis by preventing the activation of cyclin B1/CDC2 complexes. It can form a ternary complex with p120(E4F) and p53 to enhance p14ARF-induced G2 cell cycle arrest in a p53-dependent manner. p14ARF also binds to E2F1 and MYC and blocks their transcriptional activator activity without affecting MYC transcriptional repression. Human p14ARF is shown to promote the proteasomal degradation of Melanoma antigen-A11 (MAGE-A11) independent of HDM2 E3 ubiquitin ligase or lysine ubiquitination. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Minges, JT et al. (2015). J. Biol. Chem. 290, 25174-25187. Rizos, H., et al. (2003). J. Biol. Chem. 278, 4981-4989).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service