Skip to Content
Merck
All Photos(13)

Documents

ZRB1406

Sigma-Aldrich

Anti-cGAS Antibody, clone 1B5 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-C6orf150, Anti-MB21D1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B5, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 58.81 kDa
observed mol wt ~59 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminus

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1B5 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Cyclic GMP-AMP synthase (cGAS). It targets an epitope within 22 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the N-terminal region of human Cyclic GMP-AMP synthase (cGAS).

Application

Quality Control Testing

Evaluated by Western Blotting in Raw264.7 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected cGAS (Cyclic GMP-AMP synthase) in Raw264.7 cell lysate.

Tested applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected cGAS in A549 and MCF-7 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected cGAS in human placenta tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound cGAS peptide with a KD of 1.0 x 10-12 in an Affinity Binding Assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected cGAS in MCF-7 cell line.

Flow Cytometry Analysis: 1 μg from a representative lot detected cGAS in one million A549 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-cGAS, clone 1B5 ZooMAb®, Cat. No. ZRB1406, is a recombinant Rabbit monoclonal antibody that targets cGAS and is tested for use in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Cyclic GMP-AMP synthase (UniProt: Q8N884; also known as EC:2.7.7.86, cGAMP synthase, cGAS, h-cGAS, 2′3′-cGAMP synthase, Mab-21 domain-containing protein 1) is encoded by the CGAS (also known as C6orf150, MB21D1) gene (Gene ID: 115004) in human. cGAS is a nucleotidyltransferase that catalyzes the formation of cyclic GMP-AMP (cGAMP) from ATP and GTP and plays a key role in innate immunity. In resting conditions, it is localized at the cell membrane as a peripheral membrane protein by binding to phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2). Membrane localization is reported to be essential to limit the recognition of self-DNA. Upon detection of double-stranded DNA (dsDNA), it is released from the cell membrane into the cytosol to commence a signaling process. Its phosphorylation at tyrosine 215 by B-lymphocyte kinase (BLK) is reported to promote its cytosolic retention. Following dephosphorylation it translocates to the nucleus. cGAS can undergo acetylation at lysine 384, 394, and 414, which inhibits its activity. It acts as a key cytosolic DNA sensor. It binds to cytosolic DNA directly, leading to activation and synthesis of cGAMP, a second messenger that binds to and activates TMEM173/STING, thereby triggering type-I interferon production. It can also act as an innate immune sensor of infection by retroviruses, such as HIV-1, by detecting the presence of reverse-transcribed DNA in the cytosol and can also detect the presence of DNA from bacteria, such as Mycobaterium tuberculosis. In addition to its antiviral activity, it is also involved in the response to cellular stresses, such as senescence, DNA damage or genome instability. It acts as a suppressor of DNA repair in response to DNA damage and translocates to the nucleus following dephosphorylation at tyrosine 215 and inhibit homologous recombination repair by interacting with PARP1. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service