Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1195

Sigma-Aldrich

Anti-TNFβ/LTA Antibody, clone 3B20-H2 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CZK 4,420.00
4 X 25 μL
CZK 9,010.00

CZK 4,420.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
CZK 4,420.00
4 X 25 μL
CZK 9,010.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

CZK 4,420.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B20-H2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 22.30 kDa
observed mol wt ~17 kDa

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3B20-H2 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects TNFβ/LTA.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal half of human TNFβ (Lymphotoxin-a).

Application

Quality Control Testing

Evaluated by Western Blotting with human recombinant TNFβ/LTA.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected recombinant human TNFβ/LTA.

Tested applications

Flow Cytometry Analysis: 1 mg from a representative lot detected TNFβ/LTA in one million HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TNFβ/LTA in human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-TNFβ/LTA, clone 3B20-H2 ZooMAb®, Cat. No. ZRB1195, is Rabbit monoclonal antibody that specifically targets TNFβ/LTA and is tested for use in Flow Cytometry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Lymphotoxin-a (UniProt: P01374; also known as LT-alpha, TNF-beta, Tumor necrosis factor ligand superfamily member 1) is encoded by the LTA (also known as TNFB, TNFSF1) gene (Gene ID: 4049) in human. Lymphotoxin-a is cytokine of the tumor necrosis family that is synthesized by lymphocytes with a signal peptide (aa 1-34), which is subsequently cleaved off to generate the mature form. It is shown to be cytotoxic for a wide range of tumor cells in vitro and in vivo. It can exist as a secreted homotrimeric form or can form a heterotrimer with lymphotoxin-a to become a membrane associated form. The homotrimeric form is shown to bind to TNFRSF1A/TNFR1, TNFRSF1B/TNFBR and TNFRSF14/HVEM and the heterotrimer binds exclusively to TNFRSF3/Lymphotoxin- receptor (LTBR). Lymphotoxin- is reported to be involved in the formation of secondary lymphoid organs during development and play a role in apoptosis. Mutations in LTA gene are known to cause Psoriatic arthritis, an inflammatory seronegative arthritis, which is a heterogeneous disorder that can range from a mild, non-destructive disease to a severe, progressive, erosive arthropathy. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service