Z769363
Branson CPX series ultrasonic bath with digital timer
bath volume 1.9 L, AC/DC input 120 V AC
Synonym(s):
ultrasonic bath sonicator, ultrasonic cleaning bath
All Photos(1)
About This Item
Recommended Products
feature
model CPX1800
packaging
pack of 1 ea
manufacturer/tradename
Branson CPX-952-119R
bath size
150 mm (6 in.) × 140 mm (5.5 in.) × 100 mm (4 in.)
bath volume
1.9 L
General description
With the Bransonic® CPX, we′ve gone digital and defined a new category in ultrasonic baths. We′ve reimagined our iconic line of ultrasonic baths for total control with a fully digital Tower Control Panel. The CPX also features our new self-adaptive technology that actively tracks and adapts to changes within the bath.
All Bransonic® baths can also be used for cell separation, sample preparation and degassing of liquids.
Features:
Note: Parts should not be allowed to rest on tank bottom, see accessories for options.
All Bransonic® baths can also be used for cell separation, sample preparation and degassing of liquids.
Features:
- Degassing - can be set up to 99 minutes
- Timer - can be set to 99 minutes or run continuously
- Self-Adaptive technology
- Digital tower control panel
- Sleep mode
- All 220-230 V units meet CE standards
Note: Parts should not be allowed to rest on tank bottom, see accessories for options.
Legal Information
Bransonic is a registered trademark of Branson Ultrasonics Corp.