Skip to Content
Merck
All Photos(2)

Documents

Z757489

Sartorius Proline® Plus multichannel mechanical pippettor

12-channel, volume 10-100 μL

Synonym(s):

Multichannel pipettor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

12-channel
electronic: no

packaging

pack of 1 ea

manufacturer/tradename

Sartorius 728230

volume

10-100 μL

Looking for similar products? Visit Product Comparison Guide

General description

The Proline® Plus 12 channel mechanical pipettes offer comfort and quality for everyday manual pipetting. It incorporates durable construction with ease and lightness of use. The multichannel pipette consists of Optiload spring-loaded tip cones that are ideal for loading and ejecting the tips. It also seals the tips on all channels equally with minimal force.

Features and Benefits

  • The click-stop system enables easy volume adjustment.
  • Broad volume display for better reading.
  • Color-coded for easy matching of fitting tips
  • High chemical and UV-resistance materials to ensure a long lifetime of the pipette.
  • Fully autoclavable and requires no disassembly.
  • Easy cleaning and maintenance with only three parts to disassemble.
  • Safe-cone filters provided for models >10μL.
  • Εrgοnomic finger support reduces the grip force required to hold the pipette.
  • Range of volume-3μL (5μL for fixed) to 10mL
  • Low pipetting forces
  • Wide range of adjustable single and multi-channel models and fixed single-channel models

Legal Information

Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service