Skip to Content
Merck
All Photos(1)

Key Documents

Z744260

Wilmad® Suprasil® Low Temperature Aqueous Cell, Spectroscopic Quality

volume 100 μL, pack of 1 ea

Synonym(s):

EPR aqueous cell, aqueous cell

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

packaging

pack of 1 ea

manufacturer/tradename

Wilmad WG-808-S-Q

volume

100 μL

General description

Small Volume TE102 Variable Temperature Aqueous Cell works with most common Variable Temperature Dewar Inserts. This cell has optically polished flat windows that are joined to the polished wall ends via a special thermal fusion technique to form an ultra high precision bore flat cell. The two windows are parallel to each other within several visible light wave numbers. These items are recommended for EPR experiments that requires critical coupling.

Application

EPR

Legal Information

Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service