Z743334
Cryo-Babies® Labels
green, L × W 1.28 in. × 0.50 in., pkg of 1000 roll Labels
Synonym(s):
cryogenic vial labels, freezer vial labels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 1000 roll Labels
manufacturer/tradename
Diversified Biotech CRY-1000-G
technique(s)
cryopreservation: suitable
L × W
1.28 in. × 0.50 in.
color
green
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Pre-cut chemically inert vinyl labels withstand boiling water, conventional freezing, and liquid nitrogen storage without peeling or cracking. Labels will accept any marking instrument, and easily peel off backing sheet for application to vial. Size is 1.28 × 0.5 in., fitting precisely around standard cryogenic vials, as well as 1.5 to 2 mL microcentrifuge tubes. Cryo-Babies also withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100 °C and dry heat up to 150 °C.
Legal Information
Cryo-Babies is a registered trademark of Diversified Biotech
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service