Documents
Z741653
Sartorius filter pipette tips
volume range 0.1-10 μL, rack of, sterile
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
material
colorless
polypropylene (Virgin)
sterility
sterile
feature
DNase free
RNase free
barrier
endotoxin free
filter
packaging
pack of 960 ea (Racked)
rack of
manufacturer/tradename
Sartorius 783201
maximum volume
10 μL
volume range
0.1-10 μL
binding type
low binding surface
General description
The Sartorius filter tips of the Biohitfamily are made of polypropylene and feature a polyethylene filter barrier to prevent aerosol and liquid contamination. This filter helps protect against the rist of cross-contamination and reduces maintenance requirements of the pipette. The filter tips are suited for molecular biology, microbiology, cell culture and radioactive work.
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
  • Pipette foaming liquids like buffers and proteins
  • Pipette solvents
  • Reverse pipetting
  • Multiple dispensing functions of electronic pipettes
Other Notes
Browse all pipette tips