Key Documents
Z722804
Hettich® ROTOFIX 32A/Universal 320/320R centrifuge accessories
Rotor, 8 place without tube holders, for ROTOFIX 32A and UNIVERSAL 320
Select a Size
1 EA
CZK 12,200.00
CZK 12,200.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
CZK 12,200.00
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
CZK 12,200.00
Please contact Customer Service for Availability
feature
to hold 8 x 1467, 1468 buckets
packaging
pack of 1 ea
manufacturer/tradename
Hettich 1418
parameter
3,305 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Rotafix 32A and Universal 320/320R
Related Categories
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)