Key Documents
Z718564
Sigma-Aldrich
Molecular Cloning: A Laboratory Manual Third Edition
About This Item
ISBN-10:
0-87969-576-5
ISBN-13:
978-0-87969-576-7
Pricing and availability is not currently available.
publication info
J. Sambrook & D. Russell, Cold Spring Harbor Laboratory Press, 2000, 2344 pp., set
availability
available only in EU
not available in USA and Canada, ROW
General description
The first two editions of this manual have been mainstays of molecular biology for nearly twenty years, with an unrivalled reputation for reliability, accuracy, and clarity. In this new edition, authors Joseph Sambrook and David Russell have completely updated the book, revising every protocol and adding a mass of new material, to broaden its scope and maintain its unbeatable value for studies in genetics, molecular cell biology, developmental biology, microbiology, neuroscience, and immunology. Handsomely redesigned and presented in new bindings of proven durability, this three-volume work is essential for everyone using today′s biomolecular techniques. The opening chapters describe essential techniques, some well-established, some new, that are used every day in the best laboratories for isolating, analyzing and cloning DNA molecules, both large and small. These are followed by chapters on cDNA cloning and exon trapping, amplification of DNA, generation and use of nucleic acid probes, mutagenesis, and DNA sequencing. The concluding chapters deal with methods to screen expression libraries, express cloned genes in both prokaryotes and eukaryotic cells, analyze transcripts and proteins, and detect protein-protein interactions. The Appendix is a compendium of reagents, vectors, media, technical suppliers, kits, electronic resources and other essential information. As in earlier editions, this is the only manual that explains how to achieve success in cloning and provides a wealth of information about why techniques work, how they were first developed, and how they have evolved.
Table of Contents
Chapter 1. Plasmids and Their Usefulness in Molecular Cloning
Chapter 2. Bacteriophage lambda and Its Vectors
Chapter 3. Working with Bacteriophage M13 Vectors
Chapter 4. Working with High-capacity Vectors
Chapter 5. Gel Electrophoresis of DNA and Pulsed-field Agarose Gel Electrophoresis
Chapter 6. Preparation and Analysis of Eukaryotic Genomic DNA
Chapter 7. Extraction, Purification, and Analysis of mRNA from Eukaryotic Cells
Chapter 8. In vitro Amplification of DNA by the Polymerase Chain Reaction
Chapter 9. Preparation of Radiolabeled DNA and RNA Probes
Chapter 10. Working with Synthetic Oligonucleotide Probes
Chapter 11. Preparation of cDNA Libraries and Gene Identification
Chapter 12. DNA Sequencing
Chapter 13. Mutagenesis
Chapter 14. Screening Expression Libraries
Chapter 15. Expression of Cloned Genes in Escherichia coli
Chapter 16. Introducing Cloned Genes into Cultured Mammalian Cells
Chapter 17. Analysis of Gene Expression in Cultured Mammalian Cells
Chapter 18. Protein Interaction Technologies
Appendices
Choose from one of the most recent versions:
Certificates of Analysis (COA)