Key Documents
Z659363
Bandelin SONOPULS ultrasonic homogenizer accessories
cooling vessel KG 5 for use with HD 2200 homogenizer
Synonym(s):
sonication vessel
Select a Size
1 EA
CZK 6,420.00
CZK 6,420.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
CZK 6,420.00
About This Item
UNSPSC Code:
41101520
NACRES:
NB.74
CZK 6,420.00
Please contact Customer Service for Availability
material
glass
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 481
availability
available only in EU
volume × I.D. × H
80 mL × 35 mm × 95 mm
General description
Cooling vessel KG for sonication of temperature-sensitive samples. The cooling jacket allows temperature control using liquid coolants during sonication.
Features and Benefits
  • Simultaneous sound and cooling
  • Optimal process observation
Choose from one of the most recent versions:
Certificates of Analysis (COA)