Key Documents
Z659282
Bandelin SONOPULS ultrasonic homogenizer accessories
Rosett cell RZ 1
Synonym(s):
Glass sample vessel
Select a Size
1 EA
CZK 9,380.00
CZK 9,380.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
CZK 9,380.00
About This Item
UNSPSC Code:
41101520
NACRES:
NB.74
CZK 9,380.00
Please contact Customer Service for Availability
material
glass
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 3606
availability
available only in EU
volume × I.D. × H
25 mL × 30 mm × 85 mm
General description
A rosette cell (RZ) enables intensive and uniform sonication of liquid media. The round shape of the side arms leads to an intensified circulation of the sample.
Features and Benefits
  • Optimal fluid circulation.
  • Good heat dissipation.
  • Homogeneous sound reinforcement results
Choose from one of the most recent versions:
Certificates of Analysis (COA)