Skip to Content
Merck
All Photos(3)

Key Documents

WH0286749M1

Sigma-Aldrich

Monoclonal Anti-SALF antibody produced in mouse

clone 5F12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-stoned B/TFIIA-alpha/beta-like factor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 12,800.00

CZK 12,800.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
CZK 12,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CZK 12,800.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The STON1-GTF2A1L mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring STON1 and GTF2A1L genes. This rare transcript encodes a fusion protein composed of greater than 95% each of the individual elements, stonin 1 and general transcription factor IIA, 1-like. The significance of this co-transcribed mRNA and the function of its protein product have not yet been determined. (provided by RefSeq)

Immunogen

SALF (NP_758515, 141 a.a. ~ 249 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
SCTHPTPKVGLPDEVNPQQAESLGFQSDDLPQFQYFREDCAFSSPFWKDEGSDSHFTLDPPGSKKMFSSRNKEMPIDQKSLNKCSLNYICEKLEHLQSAENQDSLRSLS

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service