Skip to Content
Merck

V3265

VU0029767

≥98% (HPLC)

Synonym(s):

(E)-2-(4-Ethoxyphenylamino)-N′-((2-hydroxynaphthalen-1-yl)methylene)acetohydrazide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H21N3O3
CAS Number:
Molecular Weight:
363.41
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

gray-brown

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

CCOc1ccc(NCC(=O)N\N=C\c2c(O)ccc3ccccc23)cc1

InChI

1S/C21H21N3O3/c1-2-27-17-10-8-16(9-11-17)22-14-21(26)24-23-13-19-18-6-4-3-5-15(18)7-12-20(19)25/h3-13,22,25H,2,14H2,1H3,(H,24,26)/b23-13+

InChI key

JWRJGCUSLSJMMD-YDZHTSKRSA-N

Biochem/physiol Actions

VU0029767 is a selective, positive allosteric modulator at the muscarinic M1 acetylcholine receptor. It shifts concentration-dependent response to acetylcholine at least five-fold, but does not compete at the ACh binding site. VU0029767 also potentiates the effects of allosteric agonist TBPB.
VU0029767 is a selective, positive allosteric modulator at the muscarinic M1 acetylcholine receptor. VU0029767 also potentiates the effects of allosteric agonist TBPB.


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library