Skip to Content
Merck

V1889

VER-3323 hemifumarate salt

>98% (HPLC), solid

Synonym(s):

(S)-1-(6-Bromo-2,3-dihydroindol-1-yl)-2-propylamine hemifumarate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H15N2Br · 0.5C4H4O4
Molecular Weight:
313.19
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>98% (HPLC)

form

solid

color

white

solubility

H2O: soluble 9 mg/mL

SMILES string

OC(=O)\C=C\C(O)=O.C[C@H](N)CN1CCc2ccc(Br)cc12.C[C@H](N)CN3CCc4ccc(Br)cc34

Gene Information

Biochem/physiol Actions

5-HT2C/ 5-HT2B serotonin receptor agonist.

Legal Information

Manufactured and sold under exclusive license with Vernalis Group plc.

Disclaimer

Protect from light.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library