Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H36O
CAS Number:
Molecular Weight:
268.48
UNSPSC Code:
12352200
MDL number:
assay
~99% (capillary GC)
storage temp.
−20°C
SMILES string
CCCCCC\C=C/CCCCCCCCCCO
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.