Skip to Content
Merck

UC455

3-Cyano-7-ethoxycoumarin

≥97% (HPLC)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C12H9NO3
CAS Number:
Molecular Weight:
215.20
UNSPSC Code:
12161501
PubChem Substance ID:
MDL number:
Assay:
≥97% (HPLC)
Form:
solid

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C12H9NO3/c1-2-15-10-4-3-8-5-9(7-13)12(14)16-11(8)6-10/h3-6H,2H2,1H3

SMILES string

CCOc1ccc2C=C(C#N)C(=O)Oc2c1

InChI key

YAFGHMIAFYQSCF-UHFFFAOYSA-N

assay

≥97% (HPLC)

form

solid

color

yellow to orange

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

3-Cyano-7-ethoxycoumarin is a fluorescent substrate for Cytochrome P450 1A1 (CYP1A1), Cytochrome P450 1A2 (CYP1A2), Cytochrome P450 2C9 (CYP2C9) and Cytochrome P450 2C19 (CYP2C19).[1]

Application

3-Cyano-7-ethoxycoumarin has been used as a substrate in cytochrome P (CYP) inhibition for fluorescence measurement.[2]

Biochem/physiol Actions

Fluorescent probe useful in microsomal dealkylase studies.
Fluorescent probe useful in microsomal dealkylase studies. Typical drug-drug interactions resulting from enzyme inhibition.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

3-Cyano-7-ethoxycoumarin is soluble in DMSO.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid determination of enzyme activities of recombinant human cytochromes P450, human liver microsomes and hepatocytes
Ghosal A, et al.
Biopharmaceutics & Drug Disposition, 24(9), 375-384 (2003)
Development of a novel lead that targets M. tuberculosis polyketide synthase 13
Aggarwal A, et al.
Cell, 170(2), 249-259 (2017)
Anima Ghosal et al.
Biopharmaceutics & drug disposition, 24(9), 375-384 (2003-12-23)
Cytochrome P450 (CYP) substrates that yield fluorescent metabolites were used for rapid screening of drug metabolism activities of 13 recombinant human cytochromes P450, human liver microsomes and human hepatocytes. Reproducible results were obtained using a fluorescent plate reader (CytoFluor) more
Anup Aggarwal et al.
Cell, 170(2), 249-259 (2017-07-04)
Widespread resistance to first-line TB drugs is a major problem that will likely only be resolved through the development of new drugs with novel mechanisms of action. We have used structure-guided methods to develop a lead molecule that targets the
Abeer ElKhoely et al.
Journal of natural medicines, 69(3), 313-323 (2015-03-23)
Liver cancer is the fifth commonest malignancy worldwide and the third leading cause of death. Identifying novel curative and preventive therapy may improve its prognosis. In this study, thymoquinone (TQ), the most active biological ingredient of Nigella sativa Linn, was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service