Skip to Content
Merck

UC432

AAMU

Synonym(s):

5-Acetylamino-6-amino-3-methyluracil, N-(4-Amino-1,2,3,6-tetrahydro-1-methyl-2,6-dioxo-5-pyrimidinyl)acetimide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H10N4O3
CAS Number:
Molecular Weight:
198.18
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12161501
MDL number:
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

color

white to light pink

mp

≥305 °C

storage temp.

2-8°C

SMILES string

CN1C(=O)NC(N)=C(NC(C)=O)C1=O

InChI

1S/C7H10N4O3/c1-3(12)9-4-5(8)10-7(14)11(2)6(4)13/h8H2,1-2H3,(H,9,12)(H,10,14)

InChI key

POQOTWQIYYNXAT-UHFFFAOYSA-N

Application

CYP1A2 metabolite of caffeine

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library