Skip to Content
Merck
All Photos(1)

Key Documents

UC282

Sigma-Aldrich

6β-Hydroxytestosterone

≥97% (HPLC)

Synonym(s):

4-Androstene-6β,17β-diol-3-one, 6β, 17β,-Dihydroxyandrost-4-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O3
CAS Number:
Molecular Weight:
304.42
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:

Assay

≥97% (HPLC)

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white to off-white

mp

≥205 °C

solubility

acetonitrile: soluble
ethanol: soluble
methanol: soluble

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](C[C@@H](O)C4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O

InChI

1S/C19H28O3/c1-18-7-5-11(20)9-15(18)16(21)10-12-13-3-4-17(22)19(13,2)8-6-14(12)18/h9,12-14,16-17,21-22H,3-8,10H2,1-2H3/t12-,13-,14-,16+,17-,18+,19-/m0/s1

InChI key

XSEGWEUVSZRCBC-ZVBLRVHNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6β-Hydroxytestosterone has been used for CYP3A activity assays in rat hepatocytes and has also been used for CYP3A4 substrate selection studies.

Biochem/physiol Actions

6β-Hydroxytestosterone is a CYP3A4 metabolite; androgenic.

Preparation Note

6β-Hydroxytestosterone is soluble in methanol, acetonitrile and ethanol.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra Galetin et al.
The Journal of pharmacology and experimental therapeutics, 314(1), 180-190 (2005-03-24)
The complexity of in vitro kinetic phenomena observed for CYP3A4 substrates (homo- or heterotropic cooperativity) confounds the prediction of drug-drug interactions, and an evaluation of alternative and/or pragmatic approaches and substrates is needed. The current study focused on the utility
Yoshitaka Miyamoto et al.
Cell transplantation, 18(5), 677-681 (2009-09-25)
Three-dimensional culture procedures have attracted attention in various fields of cell biology. A newly developed cell array assisted in the formation of hepatocyte spheroids by two innovations: 1) micropatterning by a hydrophilic polymer, and 2) the use of bovine carotid
Yong Liu et al.
Biological & pharmaceutical bulletin, 27(10), 1555-1560 (2004-10-07)
The intestinal bacterial metabolites of ginsenosides are responsible for the main pharmacological activities of ginseng. The purpose of this study was to find whether these metabolites influence hepatic metabolic enzymes and to predict the potential for ginseng-prescription drug interactions. Utilizing
Zhen Lou et al.
The Journal of steroid biochemistry and molecular biology, 82(4-5), 413-424 (2003-02-19)
Intestinal or hepatic microsomes from channel catfish converted [4-14C]-testosterone to three major metabolites: 6 beta-hydroxytestosterone, androstenedione and a third metabolite. Formation of the unknown metabolite required NADPH as cofactor. When incubated with 200 microM testosterone, the rate of formation of
Dan A Rock et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(12), 2410-2413 (2008-09-04)
Cytochrome P450 (P450) reaction phenotyping is a key process toward accurately determining the contribution of different P450s to the metabolism of new chemical entities. The significance of P450s to drug disposition has led to the identification of selective chemical and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service