Skip to Content
Merck

Skip To

UC169

Hydroxybufuralol maleate salt

≥98% (HPLC), Dufuralol metabolite, solid

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C16H23NO3 · C4H4O4
Molecular Weight:
393.43
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid


Product Name

Hydroxybufuralol maleate salt, ≥98% (HPLC)

assay

≥98% (HPLC)

form

solid

color

white to off-white

mp

159-160.5 °C

solubility

H2O: soluble

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.CC(O)c1cccc2cc(oc12)C(O)CNC(C)(C)C

InChI

1S/C16H23NO3.C4H4O4/c1-10(18)12-7-5-6-11-8-14(20-15(11)12)13(19)9-17-16(2,3)4;5-3(6)1-2-4(7)8/h5-8,10,13,17-19H,9H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

UCHVLIWUAUXBBC-BTJKTKAUSA-N

Application

Hydroxybufuralol maleate salt can be used for metabolic assays of bufuralol.

Biochem/physiol Actions

Metabolite of bufuralol; product of microsomal oxidation

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

Hydroxybufuralol maleate salt is soluble in water.


Still not finding the right product?

Explore all of our products under Hydroxybufuralol maleate salt


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters