Skip to Content
Merck
All Photos(1)

Key Documents

TV0010

Sigma-Aldrich

pACD4 TargeTron Vector Set

No selectable marker.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

shipped in

wet ice

storage temp.

−20°C

General description

The pACD4 vector set (no selectable marker) is a set of four linearized vectors intended for use with the TargeTron Gene Knockout System (TA0100) when the use of a selectable marker is not desired. Each vector in the pACD4 vector set is similar to the pACD4K-C TargeTron vector with the main difference being that they do not have a kanamycin RAM marker to select for chromosomal insertion. The pACD4 vector set also contains either an A, C, G, or T at the δ + 1 position. This is to facilitate proper base pairing of group II intron precursor RNA for different target site designs to allow for more efficient precursor RNA splicing to form active TargeTron RNPs.

Other Notes

For more information and to view applications data, please visit www.sigma-aldrich.com/targetron.

Legal Information

TargeTron is a trademark of InGex, LLC

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service